Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-Valinamide hydrochloride 98%

Supplier: AMBEED, INC
Warning

A121602-1G A121602-10G A121602-250MG A121602-25G A121602-1G A121602-100G
103800-092EA 19 USD InStock
103800-092 76797-410 76797-412 76797-414 77312-220 77878-899
D-Valinamide hydrochloride 98%
D-Valinamide hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₃ClN₂O
MW: 152.6225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00270619
CAS Number: 133170-58-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty