Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Histidine methyl ester dihydrochloride 98%

Supplier: AMBEED, INC
Warning

A108670-1000G A108670-100G A108670-10G A108670-1G A108670-25G A108670-5G A108670-500G
103823-164EA 6 USD InStock
103823-164 77107-834 77107-836 77107-838 77107-840 77107-842 77283-768 77684-979
L-Histidine methyl ester dihydrochloride 98%
L-Histidine methyl ester dihydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₁N₃O₂
Melting Pt: 207 °C
Flash Pt: 176.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012701
CAS Number: 7389-87-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty