1-Isoquinolinecarboxylic acid 98%

Supplier: AMBEED, INC
Warning

A151350-5G A151350-100G A151350-10G A151350-500G A151350-25G A151350-5G
103842-400EA 11 USD
103842-400 77272-992 77272-994 77272-996 77317-948 77317-950
1-Isoquinolinecarboxylic acid 98%
1-Isoquinolinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 345
Melting Pt: 161
Storage Temperature: Ambient
MDL Number: MFCD00006901
CAS Number: 486-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR