Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

13-cis-Retinoic acid 98% (by HPLC)

Supplier: AMBEED, INC
Danger

A181906-1G A181906-5G A181906-1G A181906-5G
103807-666EA 8 USD InStock
103807-666 103846-036 77301-754 77301-756
13-cis-Retinoic acid 98% (by HPLC)
13-cis-Retinoic acid
Precaución: for Research Use Only


Formula: C₂₀H₂₈O₂
Boiling Pt: 462.8 °C (760 mm Hg)
Melting Pt: 174…177 °C
Flash Pt: 350.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00079542
CAS Number: 4759-48-2
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty