Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Trimethylbenzylamine 97%

Supplier: AMBEED, INC
Warning

A348984-1G A348984-5G A348984-10G A348984-25G
103826-006EA 11 USD InStock
103826-006 103836-996 76840-078 76840-080 77251-500 77251-502
2,4,6-Trimethylbenzylamine 97%
2,4,6-Trimethylbenzylamine
Precaución: for Research Use Only


Formula: C₁₀H₁₅N
Boiling Pt: 246.5 °C (760 mm Hg)
Flash Pt: 102.4 °C
Storage Temperature: Ambient
MDL Number: MFCD03411013
CAS Number: 40393-99-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty