Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2,6-dichloroisonicotinate 95%

Supplier: AMBEED, INC
Warning

A105045-1G A105045-5G A105045-100G A105045-100MG A105045-10G A105045-250MG A105045-25G A105045-1G A105045-5G
103796-822EA 8 USD InStock
103796-822 103822-550 76796-454 76796-456 76796-458 76796-460 76796-462 77309-414 77309-416
Methyl-2,6-dichloroisonicotinate 95%
Methyl 2,6-dichloroisonicotinate
Precaución: for Research Use Only


Formula: C₇H₅Cl₂NO₂
Boiling Pt: 298.3 °C (760 mm Hg)
Melting Pt: 82 °C
Flash Pt: 134 °C
Storage Temperature: Ambient
MDL Number: MFCD00044409
CAS Number: 42521-09-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty