Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-bromo-3-methylbenzoate 98%

Supplier: AMBEED, INC
Warning

A184962-1G A184962-5G A184962-10G A184962-500G A184962-25G A184962-5G A184962-1G
103815-804EA 5 USD InStock
103815-804 103846-370 76998-814 76998-816 77084-000 77084-002 77301-936
Methyl-2-bromo-3-methylbenzoate 98%
Methyl 2-bromo-3-methylbenzoate
Precaución: for Research Use Only


Formula: C₉H₉BrO₂
Boiling Pt: 267.4 °C (760 mm Hg)
Flash Pt: 115.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00144762
CAS Number: 131001-86-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty