Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-chloro-3-fluoro-4-pyridinecarboxylate 95%

Supplier: AMBEED, INC
Warning

A104658-1G A104658-5G A104658-250MG A104658-1G
103796-752EA 24 USD InStock
103796-752 103822-470 77135-644 77283-260
Methyl-2-chloro-3-fluoro-4-pyridinecarboxylate 95%
Methyl-2-chloro-3-fluoro-4-pyridinecarboxylate
Precaución: for Research Use Only


Formula: C₇H₅ClFNO₂
Boiling Pt: 257.5 °C (760 mm Hg)
Flash Pt: 109.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972402
CAS Number: 628691-95-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty