Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-oxo-1-cycloheptanecarboxylate 97%

Supplier: AMBEED, INC
Warning

A403445-5G A403445-250MG A403445-1G A403445-5G A403445-25G
103827-094EA 21 USD InStock
103827-094 103838-134 76725-106 76841-606 76841-608 77879-877
Methyl 2-oxo-1-cycloheptanecarboxylate 97%
Methyl 2-oxo-1-cycloheptanecarboxylate
Precaución: for Research Use Only


Formula: C₉H₁₄O₃
MW: 170.206 g/mol
Boiling Pt: 242.1 °C (760 mm Hg)
Flash Pt: 105.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00010809
CAS Number: 52784-32-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty