Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-(methylamino)but-2-enoate 97%

Supplier: AMBEED, INC
Warning

A509253-1G A509253-5G A509253-100MG
103828-366EA 19 USD InStock
103828-366 103839-466 76843-452 76843-454 77169-404
Methyl-3-(methylamino)but-2-enoate 97%
Methyl-3-(methylamino)but-2-enoate
Precaución: for Research Use Only


Formula: C₆H₁₁NO₂
Boiling Pt: 187.5 °C (760 mm Hg)
Melting Pt: 60…63 °C
Flash Pt: 67.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00027383
CAS Number: 13412-12-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty