Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-hydroxyisoxazole-5-carboxylate 97%

Supplier: AMBEED, INC
Warning

A125311-1G A125311-10G A125311-250MG A125311-25G A125311-1G A125311-5G
103800-666EA 16 USD InStock
103800-666 76718-696 76718-698 76718-700 77312-820 77312-822
Methyl-3-hydroxyisoxazole-5-carboxylate 97%
Methyl-3-hydroxyisoxazole-5-carboxylate
Precaución: for Research Use Only


Formula: C₅H₅NO₄
MW: 143.1000 g/mol
Melting Pt: 160…163 °C
Flash Pt: 155.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00062870
CAS Number: 10068-07-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty