Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4,5,6,7-tetrahydrothieno[3,2-c]pyridine-2-carboxylate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A378933-100MG A378933-250MG
103826-634EA 19 USD InStock
103826-634 103837-668 77264-216 77264-218
Methyl 4,5,6,7-tetrahydrothieno[3,2-c]pyridine-2-carboxylate hydrochloride 95%
Methyl 4,5,6,7-tetrahydrothieno[3,2-c]pyridine-2-carboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₂ClNO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD27991978
CAS Number: 1992989-11-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty