Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-amino-3-chlorobenzoate 98%

Supplier: AMBEED, INC
Warning

A217162-1G A217162-5G A217162-100G A217162-10G A217162-1G A217162-25G A217162-5G
103818-604EA 8 USD InStock
103818-604 103849-332 76721-850 76721-852 77087-948 77087-950 77087-952
Methyl-4-amino-3-chlorobenzoate 98%
Methyl 4-amino-3-chlorobenzoate
Precaución: for Research Use Only


Formula: C₈H₈ClNO₂
Boiling Pt: 321.6 °C (760 mm Hg)
Melting Pt: 107…111 °C
Flash Pt: 148.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01320686
CAS Number: 84228-44-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty