Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-bromothiophene-3-carboxylate 97%

Supplier: AMBEED, INC
Warning

A123389-1G A123389-100MG A123389-10G A123389-250MG A123389-5G A123389-1G
103800-396EA 24 USD InStock
103800-396 76822-544 76822-546 76822-548 76822-550 77312-514
Methyl 4-bromothiophene-3-carboxylate 97%
Methyl 4-bromothiophene-3-carboxylate
Precaución: for Research Use Only


Formula: C₆H₅BrO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD12913867
CAS Number: 78071-37-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty