Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-amino-3,5-dibromopicolinate 95%

Supplier: AMBEED, INC
Warning

A415712-1G A415712-25G A415712-5G A415712-100MG A415712-10G A415712-250MG
103827-298EA 25 USD InStock
103827-298 103838-342 76841-926 76841-928 76841-930 76877-038 76877-040 76877-042
Methyl-6-amino-3,5-dibromopicolinate 95%
Methyl 6-amino-3,5-dibromopicolinate
Precaución: for Research Use Only


Formula: C₇H₆Br₂N₂O₂
Boiling Pt: 366 °C (760 mm Hg)
Flash Pt: 175.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13195771
CAS Number: 443956-21-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty