Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[(Methylamino)methyl]tetrahydro-2H-pyran 96%

Supplier: AMBEED, INC
Warning

A254724-1G A254724-5G A254724-1G A254724-5G A254724-250MG
103812-158EA 103 USD InStock
103812-158 103852-010 77091-454 77091-456 77166-058
4-[(Methylamino)methyl]tetrahydro-2H-pyran 96%
4-[(Methylamino)methyl]tetrahydro-2H-pyran
Precaución: for Research Use Only


Formula: C₇H₁₅NO
Boiling Pt: 110 °C (15 mm Hg)
Flash Pt: 65.056 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06739009
CAS Number: 439081-52-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty