Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl hydrogen adipate 98%

Supplier: AMBEED, INC
Warning

A277606-25G A277606-100G A277606-10G A277606-500G A277606-1000G
103853-292EA 5 USD InStock
103853-292 77093-194 77113-990 77113-992 77113-994 77685-782
Methyl hydrogen adipate 98%
Methyl hydrogen adipate
Precaución: for Research Use Only


Formula: C₇H₁₂O₄
Melting Pt: 7…9 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004418
CAS Number: 627-91-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty