Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentanimidamide hydrochloride 98%

Supplier: AMBEED, INC
Warning

A478125-25G A478125-5G A478125-100G A478125-1G
103839-170EA 36 USD InStock
103839-170 76843-104 76843-106 76877-804 76877-806
Pentanimidamide hydrochloride 98%
Pentanimidamide hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₃ClN₂
Storage Temperature: Ambient
MDL Number: MFCD11052491
CAS Number: 18257-46-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty