Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium di-tert-butyl phosphate 98%

Supplier: AMBEED, INC
Warning

A101378-1G A101378-100G A101378-10G A101378-25G A101378-1G A101378-5G
103795-940EA 5 USD InStock
103795-940 76796-272 76796-274 76796-276 77297-076 77878-793
Potassium di-tert-butyl phosphate 98%
Potassium di-tert-butyl phosphate
Precaución: for Research Use Only


Formula: C₈H₁₈KO₄P
Melting Pt: 247…252 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03840344
CAS Number: 33494-80-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty