S-Acetyl-L-glutathione 95%

Supplier: AMBEED, INC
Warning

A265161-1G A265161-10G A265161-250MG A265161-100G
103812-712EA 7 USD
103812-712 103852-606 76722-908 76722-910 77601-136
S-Acetyl-L-glutathione 95%
S-Acetyl-L-glutathione

Precaución: for Research Use Only

Formula: C₁₂H₁₉N₃O₇S
MW: 349.36 g/mol
Boiling Pt: 770.2 °C (760 mm Hg)
Flash Pt: 419.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09952588
CAS Number: 3054-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR