Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 3,6-diazabicyclo[3.1.0]hexane-3-carboxylate 97%

Supplier: AMBEED, INC
Warning

A275459-1G A275459-100MG A275459-250MG
103813-258EA 403 USD InStock
103813-258 76988-386 76988-388
tert-Butyl 3,6-diazabicyclo[3.1.0]hexane-3-carboxylate 97%
tert-Butyl 3,6-diazabicyclo[3.1.0]hexane-3-carboxylate
Precaución: for Research Use Only


Formula: C₉H₁₆N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD18909272
CAS Number: 1262407-18-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty