Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,5,6-Tetramethylpyrazine 95%

Supplier: AMBEED, INC
Warning

A273845-5G A273845-25G A273845-5G A273845-10G A273845-500G
103853-086EA 5 USD InStock
103853-086 77092-894 77092-896 77153-674 77685-775
2,3,5,6-Tetramethylpyrazine 95%
2,3,5,6-Tetramethylpyrazine
Precaución: for Research Use Only


Formula: C₈H₁₂N₂
Melting Pt: 77…80 °C
Flash Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00006146
CAS Number: 1124-11-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty