Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A207006-5G A207006-1G A207006-5G A207006-250MG A207006-25G
103817-752EA 6 USD InStock
103817-752 103848-432 77086-694 77086-696 77097-576 77097-578
Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid 98%
Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid
Precaución: for Research Use Only


Formula: C₂₃H₂₅NO₄
Boiling Pt: 604.9 °C (760 mm Hg)
Melting Pt: 182…186 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00273480
CAS Number: 167690-53-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty