Cole-Parmer® Essentials Economy Narrow-Mouth Plastic Bottles, PPCO, Antylia Scientific
Supplier: Argos Technologies
06034-26 06034-30 06034-34 06034-38 06034-44
76534-054PK 18.48 USD
76534-054 76534-056 76534-058 76534-060 76534-062
Cole-Parmer® Essentials Economy Narrow-Mouth Plastic Bottles, PPCO, Antylia Scientific
Bottles Plastic Bottles
Store chemical samples safely.
  • Leakproof, polypropylene caps
  • Built-in shoulder loop for easier handling
  • Autoclavable
Use these sturdy bottles whenever excellent chemical resistance is desired for most acids, bases, and alcohols. Ideal for storing chemicals and distilled water. Larger bottles feature built-in shoulder loop.