Human Recombinant IL-6R/CD126 Protein (ECD, Fc Tag) (from HEK293 Cells)
Supplier: wmf consumer eletric GmbH
10398-H02H-1 10398-H02H-100
103885-928EA 2280.58 USD InStock
103885-928 103885-968
Human Recombinant IL-6R/CD126 Protein (ECD, Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human IL6R (NP_000556.1) (Met1-Pro365) was expressed with the Fc region of human IgG1 at the C-terminus.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty