Human Recombinant ACE2/Angiotensin-Converting Enzyme 2 Protein (Fc Tag) (from HEK293 Cells)
Supplier: wmf consumer eletric GmbH
10108-H02H-20 10108-H02H-100
103678-330EA 300.98 USD InStock
103678-330 76537-302
Human Recombinant ACE2/Angiotensin-Converting Enzyme 2 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Ser 740) of human ACE2 precursor (NP_068576.1) was expressed with the fused Fc region of human IgG1 at the C-terminus.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty