trFluor™ Anti-IgG Goat Polyclonal Antibody [clone:IgG] trFluor™ Eu, AAT Bioquest
Supplier: AAT BIOQUEST INC
16668 16820
76482-590EA 375.27 USD
76482-590 76482-592
trFluor™ Anti-IgG Goat Polyclonal Antibody [clone:IgG] trFluor™ Eu, AAT Bioquest
Antibodies
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
  • Large stokes shift
  • Improved stability
  • High emission yield
  • Long emission half-life
Precaución: For research use only.