Screen Quest™ Membrane Potential Assay Kit, Red Fluorescence, AAT Bioquest
Supplier: AAT BIOQUEST INC
36004 36005 36006
76483-914EA 249.58 USD
76483-914 76483-916 76483-918
Screen Quest™ Membrane Potential Assay Kit, Red Fluorescence, AAT Bioquest
Assays Cellular Assays
Membrane potential is the difference in voltage between the interior and exterior of a cell.
  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Precaución: For research use only.