ReadiUse™ Rapid Luminometric ATP Assay Kit, AAT Bioquest
Supplier: AAT BIOQUEST INC
21601
21602
21603
76484-712EA
124.31
USD
76484-712
76484-714
76484-716
ReadiUse™ Rapid Luminometric ATP Assay Kit, AAT Bioquest
Assays
Cellular Assays
Adenosine triphosphate (ATP) plays a fundamental role in cellular energetics, metabolic regulation and cellular signaling.
- Extreme sensitivity
- Stable signal with >2 hour half-life
- No DTT used
Precaución: For research use only.