Potassium acetate, anhydrous 99+%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

AC22015-0010 AC22015-0050 AC22015-0250 AC22015-1000
TS22015-0010EA 90 USD
TS22015-0010 TS22015-0050 TS22015-0250 TS22015-1000
Potassium acetate, anhydrous 99+%, pure
Potassium acetate
Formula: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 304
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=2c34b38ae71852d9f28d932f9185e4f697479ff6&prid=web&clid=SAPDAM