Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bismuth(III) nitrate pentahydrate 98%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC45411-0010 AC45411-0050
TS45411-0010EA 135 USD InStock
TS45411-0010 TS45411-0050
Bismuth(III) nitrate pentahydrate 98%, pure
Bismuth(III) nitrate pentahydrate


Formula: Bi(NO₃)₃·5H₂O
Boiling Pt: 80 °C (760 mmHg)
Melting Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00149157
CAS Number: 10035-06-0
UN: 1477
ADR: 5.1,II
Order Now
Specification Test Results
Appearance White Crystals
Titration Complexometric ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty