Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium tetrachloroaurate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC31612-0010 AC31612-0050
TS31612-0010EA 157 USD InStock
TS31612-0010 TS31612-0050
Potassium tetrachloroaurate 98%
Potassium tetrachloroaurate


Formula: AuCl₄K
Melting Pt: 357 °C
Storage Temperature: Ambient
MDL Number: MFCD00043088
CAS Number: 13682-61-6
EINECS: 237-190-1
UN: 3260
Order Now
Specification Test Results
Appearance Yellow to light orange Crystalline powder
Assay ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty