3-Chloropropylamine hydrochloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC16201-0010 AC16201-0500 AC16201-2500
TS16201-0010EA 456 USD
TS16201-0010 TS16201-0500 TS16201-2500
3-Chloropropylamine hydrochloride 98%
3-Chloropropylamine hydrochloride
Formula: C₃H₈ClN
MW: 130.01 g/mol
Melting Pt: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00012913
CAS Number: 6276-54-6

Order Now

Specification Test Results

Appearance Off-white to yellow to light beige Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98 % (Ionised Chloride)
Water ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR