Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(2,4-pentanedionato)molybdenum(VI) dioxide 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

AC21114-0500 AC21114-2500
TS21114-0500EA 117 USD InStock
TS21114-0500 TS21114-2500
Bis(2,4-pentanedionato)molybdenum(VI) dioxide 97%
Bis(2,4-pentanedionato)molybdenum(VI) dioxide


Formula: C₁₀H₁₄MoO₆
Melting Pt: 184 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011506
CAS Number: 17524-05-9
Order Now
Specification Test Results
Appearance Yellow to gray-green to light brown Crystalline powder
Infrared spectrum Conforms
Elemental Analysis ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty