Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isobutyllithium 1.7 M in heptane, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC37759-1000 AC37759-8000
TS37759-1000EA 261 USD InStock
TS37759-1000 TS37759-8000
Isobutyllithium 1.7 M in heptane, AcroSeal™
Isobutyllithium


Formula: C₄H₉Li
Density: 0.7 g/cm³
Storage Temperature: Freezer
MDL Number: MFCD01863085
CAS Number: 920-36-5
UN: 3394
ADR: 4.2,
Order Now
Specification Test Results
Appearance Colorless to yellow Solution (May develop some turbidity or precipitate)
Titration Active base 1.61 to 1.83 M

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty