Acetonitrile, extra dry 99.9+%, AcroSeal™
Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
|
Danger
|
32681-0010
32681-0025
326811000
TS32681-0010EA
175
USD
TS32681-0010
TS32681-0025
TS326811000
Acetonitrile, extra dry 99.9+%, AcroSeal™
Acetonitrile
Formula:
H₃CCN MW: 41.05 g/mol Boiling Pt: 81.6 Melting Pt: -45.7 Density: 0.781 g/cm³ Flash Pt: 2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3 II REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Water | ≤0.001 % (Coulometric) |
GC | ≥99.90 % |
Residue after evaporation | ≤0.0005 % |
Acidity (CH3COOH) | ≤0.002 % |
Acetamide | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...