Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

n-Butyllithium 1.6 M in hexanes, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

18127-1000 18127-5000 18127-8000
TS18127-1000EA 77 USD InStock
TS18127-1000 TS18127-5000 TS18127-8000
n-Butyllithium 1.6 M in hexanes, AcroSeal™
n-Butyllithium


Formula: C₄H₉Li
Melting Pt: –95 °C
Density: 0.68 g/cm³
Flash Pt: –18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009414
CAS Number: 109-72-8
EINECS: 203-698-7
UN: 3394
ADR: 4.2,I
Order Now
Specification Test Results
Appearance Clear to hazy colorless to yellow Solution (May develop some turbidity or precipitate)
Titration Active base 1.49 to 1.70 M
Free base ≤1 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty