Chromeazurol S (Mordant blue 29)
Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
|
Warning
|
19005-1000
19005-0250
TS19005-1000EA
289
USD
TS19005-1000
TS19005-0250
Chromeazurol S (Mordant blue 29)
Chromeazurol S (Mordant blue 29)
Formula:
C₂₃H₁₆Cl₂O₉S MW: 605.28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00001615 CAS Number: 1667-99-8 |
Specification Test Results
Appearance | Dark brown to black Crystalline powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
UV Lambda max | 424 to 436 nm (c=0.03 g/l in H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...