Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

15656-0250 15656-1000
TS15656-0250EA 111 USD InStock
TS15656-0250 TS15656-1000
4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) 97%
4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane)


Formula: C₂₀H₁₇ClO
Melting Pt: 121…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000814
CAS Number: 14470-28-1
EINECS: 238-463-8
Order Now
Specification Test Results
Appearance Light yellow to beige to pink Crystalline powder
Infrared spectrum Conforms
Melting point 121 to 125 °C
Assay ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty