Sulphur trioxide trimethylamine complex 95%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15935-0250 15935-1000 15935-5000
TS15935-0250EA 41 USD
TS15935-0250 TS15935-1000 TS15935-5000
Sulphur trioxide trimethylamine complex 95%
Sulphur trioxide trimethylamine complex
Formula: C₃H₉NO₃S
MW: 139.17 g/mol
Melting Pt: 232 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012421
CAS Number: 3162-58-1
UN: 3261
ADR: 8 II