Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine 97+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

38996-0010 38996-0050
TS38996-0010EA 99 USD InStock
TS38996-0010 TS38996-0050
(S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine 97+%
(S)-(+)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine


Formula: C₉H₁₆N₂O₂
Density: 1.028 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00066229
CAS Number: 78342-42-4
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4600 to 1.4620 (20°C, 589 nm)
GC ≥97.0 %
Specific optical rotation +96° to +107° (20°C, 589 nm) (C=1, EtOH)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty