Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Barium diphenylamine-4-sulphonate, pure indicator grade

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

40168-0050 40168-0100 40168-0500
TS40168-0050EA 84 USD InStock
TS40168-0050 TS40168-0100 TS40168-0500
Barium diphenylamine-4-sulphonate, pure indicator grade
Barium diphenylamine-4-sulphonate


Formula: C₂₄H₂₀BaN₂O₆S₂
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007497
CAS Number: 6211-24-1
EINECS: 228-278-0
UN: 1564
ADR: 6.1,III
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c37e55aa4075d4ea7537fbef23f9195faa03f14&vid=8c37e55aa4075d4ea7537fbef23f9195faa03f14&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Off-white to grey Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (105°C)
Sensitivity (as indicator) Passes test

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty