Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aliskiren hemifumarate

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

46379-0010 46379-2500
TS46379-0010EA 490 USD InStock
TS46379-0010 TS46379-2500
Aliskiren hemifumarate
Aliskiren hemifumarate


Formula: C₃₀H₅₃N₃O₆·0.5C₄H₄O₄
Storage Temperature: Refrigerator
MDL Number: MFCD27756642
CAS Number: 173334-58-2
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH 9.3 to 9.7 % (fumaric acid dry substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty