Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Iodic acid 99.5% for analysis

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

20645-0010 20645-0500 20645-2500
TS20645-0010EA 1114 USD InStock
TS20645-0010 TS20645-0500 TS20645-2500
Iodic acid 99.5% for analysis
Iodic acid


Formula: HIO₃
Melting Pt: 110 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011348
CAS Number: 7782-68-5
EINECS: 231-962-1
UN: 3085
ADR: 5.1,II
Merck Index: 13,05034
Order Now
Specification Test Results
Appearance White to light beige Powder or crystals
Titration Iodometric ≥99.4 %
Iron (Fe) ≤30 ppm
Heavy metals (as Pb) ≤50 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty