Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trichloroacetamide 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

20292-0250 20292-1000 20292-5000
TS20292-0250EA 29 USD InStock
TS20292-0250 TS20292-1000 TS20292-5000
Trichloroacetamide 98+%
Trichloroacetamide


Formula: C₂H₂Cl₃NO
Boiling Pt: 238…240 °C (760 mmHg)
Melting Pt: 139…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00008009
CAS Number: 594-65-0
EINECS: 209-849-3
Order Now
Specification Test Results
Appearance White to off-white Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 139 to 143 °C
HPLC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty