Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethyl-1,3-pentadiene 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15536-0050 15536-0250 15536-1000
TS15536-0050EA 84 USD InStock
TS15536-0050 TS15536-0250 TS15536-1000
2,4-Dimethyl-1,3-pentadiene 98%
2,4-Dimethyl-1,3-pentadiene


Formula: C₇H₁₂
Boiling Pt: 94 °C (760 mmHg)
Melting Pt: –114 °C
Density: 0.74 g/cm³
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00008903
CAS Number: 1000-86-8
EINECS: 213-677-4
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4420 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty