1,3,5,7-Cyclooctatetraene 98% stabilised

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

16057-0025 16057-0100 16057-2500
TS16057-0025EA 142 USD
TS16057-0025 TS16057-0100 TS16057-2500
1,3,5,7-Cyclooctatetraene 98% stabilised
1,3,5,7-Cyclooctatetraene
Formula: C₈H₈
MW: 104.15 g/mol
Boiling Pt: 143
Melting Pt: -7
Density: 0.92 g/cm³
Flash Pt: 22 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004161
CAS Number: 629-20-9
EINECS: 211-080-3
UN: 2358
ADR: 3 II

Order Now

Specification Test Results

Appearance Turbid yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5365 to 1.5385 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 0.1 % hydroquinone


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR