Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3'-Dithiodipropionic acid 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

14890-0500 14890-2500
TS14890-0500EA 36 USD InStock
TS14890-0500 TS14890-2500
3,3'-Dithiodipropionic acid 98%
3,3'-Dithiodipropionic acid


Formula: C₆H₁₀O₄S₂
Melting Pt: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00002780
CAS Number: 1119-62-6
EINECS: 214-284-0
UN: 3335
ADR: 9,III
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Melting point 154 to 158 °C
Titration with NaOH ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty