Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl carbazate 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

11820-0250 11820-1000 11820-5000
TS11820-0250EA 38 USD InStock
TS11820-0250 TS11820-1000 TS11820-5000
Ethyl carbazate 97%
Ethyl carbazate


Formula: C₃H₈N₂O₂
Boiling Pt: 108…110 °C (22 mmHg)
Melting Pt: 44…48 °C
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00007595
CAS Number: 4114-31-2
EINECS: 223-903-3
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to almost white (May have a pink hue) Crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 42 to 48 °C
GC ≥96.0 %
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty