Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl (R)-(-)-3-hydroxybutyrate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

34576-0010 34576-0050
TS34576-0010EA 46 USD InStock
TS34576-0010 TS34576-0050
Ethyl (R)-(-)-3-hydroxybutyrate 98%
Ethyl (R)-(-)-3-hydroxybutyrate
Appearance: Clear colorless Liquid


Formula: C₆H₁₂O₃
Boiling Pt: 75…76 °C (12 mmHg)
Density: 1.017 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00075386
CAS Number: 24915-95-5
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4195 to 1.4215 (20°C, 589 nm)
GC ≥97.5 %
Specific optical rotation -43° to -47° (20°C, 589 nm) (c=1, CHCl3)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty